3-(5-Formyl-2-thienyl)benzonitrile

Supplier: Apollo Scientific

OR470327-5G
APOSOR470327-5GEA 1840 EUR
APOSOR470327-5G
3-(5-Formyl-2-thienyl)benzonitrile
3-(5-Formyl-2-thienyl)benzonitrile
Formula: C₁₂H₇NOS
Storage Temperature: Ambient
MDL Number: MFCD06410198
CAS Number: 886508-88-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR