N-Ethylcyclopropanamine hydrochloride

Supplier: Apollo Scientific
Warning

OR470784-1G OR470784-5G
APOSOR470784-1GEA 337 EUR
APOSOR470784-1G APOSOR470784-5G
N-Ethylcyclopropanamine hydrochloride
N-Ethylcyclopropanamine hydrochloride
Formula: C₅H₁₁N·ClH
Storage Temperature: Ambient
MDL Number: MFCD11099458
CAS Number: 672302-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR