tert-Butyl iodoacetate

Supplier: Apollo Scientific

Synonyms: tert-Butyl 2-iodoacetate

OR471032-5G
APOSOR471032-5GEA 200 EUR
APOSOR471032-5G
tert-Butyl iodoacetate
tert-Butyl iodoacetate
Formula: C₆H₁₁IO₂
MW: 242.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09833728
CAS Number: 49827-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR