2-(2-Chloro-4-nitrophenyl)acetic acid

Supplier: Apollo Scientific

OR471045-1G
APOSOR471045-1GEA 128 EUR
APOSOR471045-1G
2-(2-Chloro-4-nitrophenyl)acetic acid
2-(2-Chloro-4-nitrophenyl)acetic acid
Formula: C₈H₆ClNO₄
Storage Temperature: Ambient
MDL Number: MFCD11036396
CAS Number: 73088-11-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR