1-(4-Nitrophenyl)cyclopropanecarboxylic acid

Supplier: Apollo Scientific

OR471169-1G
APOSOR471169-1GEA 108 EUR
APOSOR471169-1G
1-(4-Nitrophenyl)cyclopropanecarboxylic acid
1-(4-Nitrophenyl)cyclopropanecarboxylic acid
Formula: C₁₀H₉NO₄
Storage Temperature: Ambient
MDL Number: MFCD07374438
CAS Number: 23348-99-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR