2-Amino-6-(aminomethyl)pyridine

Supplier: Apollo Scientific

OR471266-1G
APOSOR471266-1GEA 916 EUR
APOSOR471266-1G
2-Amino-6-(aminomethyl)pyridine
2-Amino-6-(aminomethyl)pyridine
Formula: C₆H₉N₃
MW: 123.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16547512
CAS Number: 167298-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR