7-Nitro-1,2-benzisoxazol-3(2H)-one

Supplier: Apollo Scientific

OR471334-1G
APOSOR471334-1GEA 855 EUR
APOSOR471334-1G
7-Nitro-1,2-benzisoxazol-3(2H)-one
7-Nitro-1,2-benzisoxazol-3(2H)-one
Formula: C₇H₄N₂O₄
MW: 180.118 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28663329
CAS Number: 60357-78-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR