Piperazine-2-carbonitrile dihydrochloride

Supplier: Apollo Scientific

Synonyms: 2-Cyanopiperazine dihydrochloride

OR471547-5G
APOSOR471547-5GEA 381 EUR
APOSOR471547-5G
Piperazine-2-carbonitrile dihydrochloride
Piperazine-2-carbonitrile dihydrochloride
Formula: C5H11Cl2N3
MW: 184.0700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12910518
CAS Number: 187589-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR