N-Carbobenzoxyglycine methyl ester 95%

Supplier: Apollo Scientific

OR471610-100G
APOSOR471610-100GEA 114 EUR
APOSOR471610-100G
N-Carbobenzoxyglycine methyl ester 95%
N-Carbobenzoxyglycine methyl ester
Formula: C₁₁H₁₃NO₄
Storage Temperature: Ambient
MDL Number: MFCD00043631
CAS Number: 1212-53-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR