4-Chlorobenzonitrile 99%

Supplier: Apollo Scientific
Warning

OR4722-1KG OR4722-250G OR4722-50G
APOSOR4722-1KGEA 764 EUR
APOSOR4722-1KG APOSOR4722-250G APOSOR4722-50G
4-Chlorobenzonitrile 99%
4-Chlorobenzonitrile
Formula: C₇H₄ClN
Boiling Pt: 222…224 °C
Melting Pt: 90…94 °C
Density: 1,2 g/cm³ (25 °C)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00001813
CAS Number: 623-03-0
EINECS: 210-765-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR