Ethyl benzoylacetate 95%

Supplier: Apollo Scientific
Warning

OR4787-100G OR4787-25G
APOSOR4787-100GEA 56.7 EUR
APOSOR4787-100G APOSOR4787-25G
Ethyl benzoylacetate 95%
Ethyl benzoylacetate
Formula: C₁₁H₁₂O₃
Boiling Pt: 265…270 °C
Density: 1,113 g/cm³ (25 °C)
Flash Pt: 140 °C
Storage Temperature: Ambient
MDL Number: MFCD00009196
CAS Number: 94-02-0
EINECS: 202-295-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR