Ethyl 5-amino-4-bromo-1-(4-chlorophenyl)pyrazole-3-carboxylate

Supplier: Apollo Scientific

OR480054-1G
APOSOR480054-1GEA 912 EUR
APOSOR480054-1G
Ethyl 5-amino-4-bromo-1-(4-chlorophenyl)pyrazole-3-carboxylate
Ethyl 5-amino-4-bromo-1-(4-chlorophenyl)pyrazole-3-carboxylate
Formula: C₁₂H₁₁BrClN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD23135223
CAS Number: 1427022-00-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR