1-(7-Bromonaphthalen-1-yl)cyclopropan-1-amine

Supplier: Apollo Scientific

OR480166-1G
APOSOR480166-1GEA 4540 EUR
APOSOR480166-1G
1-(7-Bromonaphthalen-1-yl)cyclopropan-1-amine
1-(7-Bromonaphthalen-1-yl)cyclopropan-1-amine
Formula: C₁₃H₁₂BrN
Storage Temperature: Ambient
MDL Number: MFCD30496538
CAS Number: 1785577-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR