1,4,5,6-Tetrahydropyrrolo[3,4-c]pyrazole dihydrochloride 95%

Supplier: Apollo Scientific

Synonyms: 1,4,5,6-Tetrahydro-pyrrolo[3,4-c]pyrazole,hydrochloride

OR480295-1G OR480295-5G
APOSOR480295-1GEA 48.2 EUR
APOSOR480295-1G APOSOR480295-5G
1,4,5,6-Tetrahydropyrrolo[3,4-c]pyrazole dihydrochloride 95%
1,4,5,6-Tetrahydropyrrolo[3,4-c]pyrazole dihydrochloride
Formula: C₅H₉Cl₂N₃
Storage Temperature: Ambient
MDL Number: MFCD08457551
CAS Number: 157327-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR