Ethyl-7-bromo-1H-indole-2-carboxylate 95%

Supplier: Apollo Scientific
Warning

OR480619-1G OR480619-5G OR480619-10G
APOSOR480619-1GEA 21.9 EUR
APOSOR480619-1G APOSOR480619-5G APOSOR480619-10G
Ethyl-7-bromo-1H-indole-2-carboxylate 95%
Ethyl-7-bromo-1H-indole-2-carboxylate
Formula: C₁₁H₁₀BrNO₂
Melting Pt: 87…88 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04972046
CAS Number: 16732-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR