1-(4-Amino-3-bromophenyl)ethanone 95%

Supplier: Apollo Scientific

Synonyms: 4-Amino-3-bromoacetophenone

OR480776-5G
APOSOR480776-5GEA 60.9 EUR
APOSOR480776-5G
1-(4-Amino-3-bromophenyl)ethanone 95%
1-(4-Amino-3-bromophenyl)ethanone
Formula: C₈H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD00272386
CAS Number: 56759-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR