2-Chloro-5-methylthiazole-4-carboxylic acid ethyl ester 95%

Supplier: Apollo Scientific

OR480827-5G
APOSOR480827-5GEA 169 EUR
APOSOR480827-5G
2-Chloro-5-methylthiazole-4-carboxylic acid ethyl ester 95%
2-Chloro-5-methylthiazole-4-carboxylic acid ethyl ester
Formula: C₇H₈ClNO₂S
Storage Temperature: Ambient
MDL Number: MFCD19443895
CAS Number: 907545-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR