2-(3-Aminophenyl)acetic acid

Supplier: Apollo Scientific
Warning

OR4815-10G OR4815-1G OR4815-25G OR4815-5G
APOSOR4815-10GEA 152 EUR
APOSOR4815-10G APOSOR4815-1G APOSOR4815-25G APOSOR4815-5G
2-(3-Aminophenyl)acetic acid
2-(3-Aminophenyl)acetic acid
Formula: C₈H₉NO₂
Melting Pt: 147…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00075058
CAS Number: 14338-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR