Uracil

Supplier: Apollo Scientific
Warning

OR4885-250G OR4885-100G OR4885-25G
APOSOR4885-250GEA 50.9 EUR
APOSOR4885-250G APOSOR4885-100G APOSOR4885-25G
Uracil
Uracil
Formula: C₄H₄N₂O₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00006016
CAS Number: 66-22-8
EINECS: 200-621-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR