DL-Tropic acid 98%

Supplier: Apollo Scientific
Warning

OR4935-100G OR4935-500G OR4935-25G
APOSOR4935-100GEA 63.8 EUR
APOSOR4935-100G APOSOR4935-500G APOSOR4935-25G
DL-Tropic acid 98%
DL-Tropic acid
Formula: C₉H₁₀O₃
Melting Pt: 116…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00004255
CAS Number: 552-63-6
EINECS: 209-020-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR