1-Oxa-9-azaspiro[5.5]undecane hydrochloride 95+%

Supplier: Apollo Scientific
Warning

OR500040-250MG
APOSOR500040-250MGEA 126 EUR
APOSOR500040-250MG
1-Oxa-9-azaspiro[5.5]undecane hydrochloride 95+%
1-Oxa-9-azaspiro[5.5]undecane hydrochloride
Formula: C₉H₁₇NO·ClH
MW: 191,698 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11036126
CAS Number: 71879-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR