3-Methylpiperidine 97%

Supplier: Apollo Scientific
Danger

OR5090-500G
APOSOR5090-500GEA 205 EUR
APOSOR5090-500G
3-Methylpiperidine 97%
3-Methylpiperidine
Formula: C₆H₁₃N
Boiling Pt: 125…156 °C
Melting Pt: –24 °C
Density: 0,845 g/cm³ (25 °C)
Flash Pt: 8 °C
Storage Temperature: Ambient
MDL Number: MFCD00005994
CAS Number: 626-56-2
EINECS: 210-953-6
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR