2-Bromo-6-methoxybenzonitrile 98%

Supplier: Apollo Scientific

OR50983-1G OR50983-25G OR50983-5G
APOSOR50983-1GEA 31.9 EUR
APOSOR50983-1G APOSOR50983-25G APOSOR50983-5G
2-Bromo-6-methoxybenzonitrile 98%
2-Bromo-6-methoxybenzonitrile
Formula: C₈H₆BrNO
Storage Temperature: Ambient
MDL Number: MFCD09834771
CAS Number: 1245647-50-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR