rac-AntPhos ≥98%

Supplier: Apollo Scientific

Synonyms: 4-(Anthracen-9-yl)-3-(t-butyl-2,3-dihydrobenzo[d][1,3]oxaphosphole, 4-(Anthracen-9-yl)-3-(tert-butyl)-2,3-dihydrobenzo[d][1,3]oxaphosphole, rac-AntPhos

OR510214-1G
APOSOR510214-1GEA 139 EUR
APOSOR510214-1G
rac-AntPhos ≥98%
rac-AntPhos
Formula: C₂₅H₂₃OP
MW: 370,423 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30748199
CAS Number: 1268693-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR