3-Chloro-2-formylpyridine

Supplier: Apollo Scientific
Warning

Synonyms: 3-Chloropyridine-2-carboxaldehyde

OR51758-1G OR51758-250MG
APOSOR51758-1GEA 66.7 EUR
APOSOR51758-1G APOSOR51758-250MG
3-Chloro-2-formylpyridine
3-Chloro-2-formylpyridine
Formula: C₆H₄ClNO
Storage Temperature: Refrigerator
MDL Number: MFCD09991855
CAS Number: 206181-90-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR