Ethyl 5-hydroxynicotinate

Supplier: Apollo Scientific
Warning

Synonyms: 3-(Ethoxycarbonyl)-5-hydroxypyridine, Ethyl 5-hydroxypyridine-3-carboxylate

OR51770-1G OR51770-250MG
APOSOR51770-1GEA 43.5 EUR
APOSOR51770-1G APOSOR51770-250MG
Ethyl 5-hydroxynicotinate
Ethyl 5-hydroxynicotinate
Formula: C₈H₉NO₃
MW: 167.162 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00510027
CAS Number: 59288-38-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR