tert-Butyl 3-aminopropanoate

Supplier: Apollo Scientific
Warning

OR51868-1G OR51868-250MG OR51868-5G
APOSOR51868-1GEA 105 EUR
APOSOR51868-1G APOSOR51868-250MG APOSOR51868-5G
tert-Butyl 3-aminopropanoate
tert-Butyl 3-aminopropanoate
Formula: C₇H₁₅NO₂
Boiling Pt: 192,3 °C
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD06212562
CAS Number: 15231-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR