Methyl-4-bromo-2-methylbenzoate

Supplier: Apollo Scientific
Warning

OR51873-1G OR51873-25G OR51873-5G
APOSOR51873-1GEA 49.3 EUR
APOSOR51873-1G APOSOR51873-25G APOSOR51873-5G
Methyl-4-bromo-2-methylbenzoate
Methyl 4-bromo-2-methylbenzoate
Formula: C₉H₉BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD09954957
CAS Number: 99548-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR