Methyl 5-amino-3-chloropyridine-2-carboxylate

Supplier: Apollo Scientific
Warning

OR52216-100MG OR52216-250MG OR52216-1G
APOSOR52216-100MGEA 84.1 EUR
APOSOR52216-100MG APOSOR52216-250MG APOSOR52216-1G
Methyl 5-amino-3-chloropyridine-2-carboxylate
Methyl 5-amino-3-chloropyridine-2-carboxylate
Formula: C₇H₇ClN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD18258038
CAS Number: 1256825-20-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR