3-Nitro-4-quinolinol 98%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Hydroxy-3-nitroquinoline, 3-Nitroquinolin-4-ol

OR52247-25G OR52247-100G
APOSOR52247-25GEA 77 EUR
APOSOR52247-25G APOSOR52247-100G
3-Nitro-4-quinolinol 98%
3-Nitro-4-quinolinol
Formula: C₉H₆N₂O₃
MW: 190.1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00277722
CAS Number: 50332-66-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR