4-(Chloromethyl)benzoyl chloride

Supplier: Apollo Scientific
Danger

OR52448-100G OR52448-25G
APOSOR52448-100GEA 197 EUR
APOSOR52448-100G APOSOR52448-25G
4-(Chloromethyl)benzoyl chloride
4-(Chloromethyl)benzoyl chloride
Formula: C₈H₆Cl₂O
Boiling Pt: 126…128 °C/8mmHg
Melting Pt: 30…32 °C
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00053224
CAS Number: 876-08-4
EINECS: 212-881-0
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR