4-Pentynylbenzene 97%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Phenyl-1-pentyne

OR5277-5G
APOSOR5277-5GEA 161 EUR
APOSOR5277-5G
4-Pentynylbenzene 97%
4-Pentynylbenzene
Formula: (C₆H₅)CH₂CH₂CH₂CCH
Boiling Pt: 94 °C/50mmHg
Density: 0,95 g/cm³ (25 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00039813
CAS Number: 1823-14-9
EINECS: 217-352-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR