1-Phenylethyl mercaptan 98%

Supplier: Apollo Scientific

OR52781-10G OR52781-1G
APOSOR52781-10GEA 292 EUR
APOSOR52781-10G APOSOR52781-1G
1-Phenylethyl mercaptan 98%
1-Phenylethyl mercaptan
Formula: C₈H₁₀S
Boiling Pt: 83…84 °C
Melting Pt: 58…59 °C
Density: 1,021 g/cm³ (25 °C)
Flash Pt: 76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00053515
CAS Number: 6263-65-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR