3-Amino-5-hydroxybenzoic acid 95%

Supplier: Apollo Scientific

OR52788-10G OR52788-5G
APOSOR52788-10GEA 290 EUR
APOSOR52788-10G APOSOR52788-5G
3-Amino-5-hydroxybenzoic acid 95%
3-Amino-5-hydroxybenzoic acid
Formula: C₇H₇NO₃
Storage Temperature: Ambient
MDL Number: MFCD06797571
CAS Number: 76045-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR