Methyl 2,6-dibromobenzoate 95%

Supplier: Apollo Scientific

OR52900-1G OR52900-250MG OR52900-5G
APOSOR52900-1GEA 241 EUR
APOSOR52900-1G APOSOR52900-250MG APOSOR52900-5G
Methyl 2,6-dibromobenzoate 95%
Methyl 2,6-dibromobenzoate
Formula: C₈H₆Br₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00234397
CAS Number: 873994-34-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR