Methyl 4-chloro-2-methyl-pyrazole-3-carboxylate ≥95%

Supplier: Apollo Scientific

OR52911-250MG OR52911-1G OR52911-5G
APOSOR52911-250MGEA 154 EUR
APOSOR52911-250MG APOSOR52911-1G APOSOR52911-5G
Methyl 4-chloro-2-methyl-pyrazole-3-carboxylate ≥95%
Methyl 4-chloro-2-methyl-pyrazole-3-carboxylate
Formula: C₆H₇ClN₂O₂
MDL Number: MFCD02253778
CAS Number: 400877-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR