2,4,6-Triisopropylbenzenesulphonyl chloride 98+%

Supplier: Apollo Scientific
Danger

OR5360-100G OR5360-25G
APOSOR5360-100GEA 58 EUR
APOSOR5360-100G APOSOR5360-25G
2,4,6-Triisopropylbenzenesulphonyl chloride 98+%
2,4,6-Triisopropylbenzenesulphonyl chloride
Formula: C₁₅H₂₃ClO₂S
Melting Pt: 94…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00007433
CAS Number: 6553-96-4
EINECS: 229-479-6
UN: 3261
ADR: 8,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR