3-Cyano-4-methylaniline

Supplier: Apollo Scientific
Warning

OR5392-25G OR5392-5G OR5392-1G
APOSOR5392-25GEA 147 EUR
APOSOR5392-25G APOSOR5392-5G APOSOR5392-1G
3-Cyano-4-methylaniline
3-Cyano-4-methylaniline
Formula: C₈H₈N₂
Boiling Pt: 110 °C/22mmHg
Melting Pt: 88…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00017608
CAS Number: 50670-64-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR