5'-Hydroxy-2'-methoxyacetophenone

Supplier: Apollo Scientific

OR54528-1G OR54528-250MG OR54528-5G
APOSOR54528-1GEA 87.6 EUR
APOSOR54528-1G APOSOR54528-250MG APOSOR54528-5G
5'-Hydroxy-2'-methoxyacetophenone
5'-Hydroxy-2'-methoxyacetophenone
Formula: C₉H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD04038299
CAS Number: 31405-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR