5-Amino-4-chloro-6-methoxy-2-methylpyrimidine

Supplier: Apollo Scientific
Warning

OR54529-100MG OR54529-1G OR54529-250MG OR54529-5G
APOSOR54529-100MGEA 35 EUR
APOSOR54529-100MG APOSOR54529-1G APOSOR54529-250MG APOSOR54529-5G
5-Amino-4-chloro-6-methoxy-2-methylpyrimidine
5-Amino-4-chloro-6-methoxy-2-methylpyrimidine
Formula: C₆H₈ClN₃O
Storage Temperature: Ambient
MDL Number: MFCD01837887
CAS Number: 88474-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR