Diethyl cyclopropane-1,2-dicarboxylate 95%

Supplier: Apollo Scientific

Synonyms: 1,2-Bis(ethoxycarbonyl)cyclopropane, Cyclopropane-1,2-dicarboxylic acid diethyl ester

OR55008-1G
APOSOR55008-1GEA 217 EUR
APOSOR55008-1G
Diethyl cyclopropane-1,2-dicarboxylate 95%
Diethyl cyclopropane-1,2-dicarboxylate
Formula: C₉H₁₄O₄
MW: 186.205 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00001286
CAS Number: 20561-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR