5-Bromohex-1-ene 96%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Bromo-1-hexene

OR55112-1G
APOSOR55112-1GEA 619 EUR
APOSOR55112-1G
5-Bromohex-1-ene 96%
5-Bromohex-1-ene
Formula: C₆H₁₁Br
MW: 163.056 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00049141
CAS Number: 4558-27-4
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR