1-Pentyne 99%

Supplier: Apollo Scientific
Danger

OR5512-100G OR5512-25G
APOSOR5512-100GEA 176 EUR
APOSOR5512-100G APOSOR5512-25G
1-Pentyne 99%
1-Pentyne
Formula: C₅H₈
Boiling Pt: 39…41 °C
Melting Pt: –106 to –105 °C
Density: 0,691 g/cm³ (25 °C)
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00009469
CAS Number: 627-19-0
EINECS: 210-987-1
UN: 3295
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR