5-Hydroxypicolinic acid 95%

Supplier: Apollo Scientific
Warning

OR5545-1G OR5545-5G OR5545-100G OR5545-25G
APOSOR5545-1GEA 21.9 EUR
APOSOR5545-1G APOSOR5545-5G APOSOR5545-100G APOSOR5545-25G
5-Hydroxypicolinic acid 95%
5-Hydroxypicolinic acid
Formula: C₆H₅NO₃
Melting Pt: 260…264 °C
Storage Temperature: Ambient
MDL Number: MFCD00661309
CAS Number: 15069-92-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR