2,4-Bis(methylsulphonyl)benzaldehyde

Supplier: Apollo Scientific
Warning

OR55491-1G OR55491-5G
APOSOR55491-1GEA 249 EUR
APOSOR55491-1G APOSOR55491-5G
2,4-Bis(methylsulphonyl)benzaldehyde
2,4-Bis(methylsulphonyl)benzaldehyde
Formula: C₉H₁₀O₅S₂
Storage Temperature: Ambient
MDL Number: MFCD28962763
CAS Number: 1845690-57-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR