N,N,N',N',N''-Pentamethyldiethylenetriamine

Supplier: Apollo Scientific
Danger

OR55651-100G OR55651-500G
APOSOR55651-100GEA 21.9 EUR
APOSOR55651-100G APOSOR55651-500G
N,N,N',N',N''-Pentamethyldiethylenetriamine
N,N,N',N',N''-Pentamethyldiethylenetriamine
Formula: C₉H₂₃N₃
Boiling Pt: 198 °C
Melting Pt: –20 °C
Density: 0,83 g/cm³ (25 °C)
Flash Pt: 77 °C
Storage Temperature: Ambient
MDL Number: MFCD00014876
CAS Number: 3030-47-5
UN: 2922
ADR: 8+6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR