Imidazole-4-carboxylic acid ethyl ester 98%

Supplier: Apollo Scientific
Warning

OR5606-1G OR5606-5G OR5606-25G
APOSOR5606-1GEA 85.8 EUR
APOSOR5606-1G APOSOR5606-5G APOSOR5606-25G
Imidazole-4-carboxylic acid ethyl ester 98%
Imidazole-4-carboxylic acid ethyl ester
Formula: C₆H₈N₂O₂
Melting Pt: 160…162 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07363812
CAS Number: 23785-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR