Piperidin-1-ylacetic acid hydrochloride

Supplier: Apollo Scientific
Warning

OR5652-1G OR5652-250MG
APOSOR5652-1GEA 29 EUR
APOSOR5652-1G APOSOR5652-250MG
Piperidin-1-ylacetic acid hydrochloride
Piperidin-1-ylacetic acid hydrochloride
Formula: C₇H₁₄ClNO₂
MW: 179.65 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01701566
CAS Number: 3235-68-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR