4-Iodo-2,5-xylenol 98%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Iodo-2,5-dimethylphenol, 2-Hydroxy-5-iodo-4-xylene, 5-Hydroxy-2-iodo-4-xylene, 2,5-Dimethyl-4-iodophenol

OR5710-1G OR5710-5G
APOSOR5710-1GEA 94.8 EUR
APOSOR5710-1G APOSOR5710-5G
4-Iodo-2,5-xylenol 98%
4-Iodo-2,5-xylenol
Formula: C₈H₉IO
Melting Pt: 95…97 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00219922
CAS Number: 114971-53-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR