6-Chloro-1-methyl-2-indoleboronic acid 96%

Supplier: Apollo Scientific
Warning

Synonyms: 6-Chloro-1-methylindole-2-boronic acid

OR59404-1G OR59404-250MG OR59404-100MG
APOSOR59404-1GEA 597 EUR
APOSOR59404-1G APOSOR59404-250MG APOSOR59404-100MG
6-Chloro-1-methyl-2-indoleboronic acid 96%
6-Chloro-1-methyl-2-indoleboronic acid
Formula: C₉H₉BClNO₂
Melting Pt: 118…120 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09878352
CAS Number: 957066-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR